Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSSKQEIMSD QRFRRVAKDP RFWEMPEKDR KVKIDKRFRA MFHDKKFKLN YAVDKRGRPI SHSTTEDLKR FYDLSDSDSN LSGEDSKALS QKKIKKKKTQ TKKEIDSKNL VEKKKETKKA NHKGSENKTD LDNSIGIKKM KTSCKFKIDS NISPKKDSKE FTQKNKKEKK NIVQHTTDSS LEEKQRTLDS GTSEIVKSPR IECSKTRREM QSVVQLIMTR DSDGYENSTD GEMCDKDALE EDSESVSEIG SDEESENEIT SVGRASGDDD GSEDDEEEDE DEEEDEDEDS EDDDKSDSGP DLARGKGNIE TSSEDEDDTA DLFPEESGFE HAWRELDKDA PRADEITRRL AVCNMDWDRL KAKDLLALFN SFKPKGGVIF SVKIYPSEFG KERMKEEQVQ GPVELLSIPE DAPEKDWTSR EKLRDYQFKR LKYYYAVVDC DSPETASKIY EDCDGLEFES SCSFIDLRFI PDDITFDDEP KDVASEVNLT AYKPKYFTSA AMGTSTVEIT WDETDHERIT MLNRKFKKEE LLDMDFQAYL ASSSEDEEEI EEELQGDDGV NVEEDGKTKK SQKDDEEQIA KYRQLLQVIQ EKEKKGKEND MEMEIKWVPG LKESAEEMVK NKLEGKDKLT PWEQFLEKKK EKKRLKRKQK ALAEEASEEE LPSDVDLNDP YFAEEVKQIG INKKSVKSAK DGTSPEEEIE IERQKAEMAL LMMDEDEDSK KHFNYNKIVE HQNLSKKKKK QLMKKKELIE DDFEVNVNDA RFQAMYTSHL FNLDPSDPNF KKTKAMEKIL EEKARQRERK EQELTQAIKK KESEIEKESQ RKSIDPALSM LIKSIKTKTE QFQARKKQKV K
May constitute a novel regulatory system for basal transcription. Negatively regulates ABT1. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABT1-associated protein; ESF1 homolog; ESF1, nucleolar pre-rRNA processing protein, homolog
Gene Aliases: ABTAP; bA526K24.1; C20orf6; ESF1; HDCMC28P
UniProt ID: (Human) Q9NX93
Entrez Gene ID: (Human) 51575
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support