Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
ESRRB (steroid hormone receptor ERR2) plays a role in maintenance of self-renewal and pluripotency of embryonic and trophoblast stem cells through different signaling pathways including FGF signaling pathway and Wnt signaling pathways. Upon FGF signaling pathway activation ESRRB interacts with KDM1A by directly binding to enhancer sites of ELF5 and EOMES and activates their transcription leading to self-renewal of trophoblast stem cells. ESSRB also regulates expression of multiple rod-specific genes and is required for survival of this cell type. Defects in the gene can cause deafness, autosomal recessive, 35 (DFNB35).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ERR beta-2; ERR-beta; ERRbeta; ERRbeta-2; Estrogen receptor-like 2; estrogen-related nuclear receptor beta; Estrogen-related receptor beta; nuclear receptor ERRB2; Nuclear receptor subfamily 3 group B member 2; orphan nuclear receptor; Steroid hormone receptor ERR2
Gene Aliases: DFNB35; ERR2; ERRb; ERRB2; ESRL2; ESRRB; NR3B2
UniProt ID: (Human) O95718
Entrez Gene ID: (Human) 2103
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support