Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Non-specific receptor for endothelin 1, 2, and 3. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AU022549; endothelin B receptor; Endothelin receptor non-selective type; endothelin receptor subtype B1; Endothelin receptor type B; endothelin-B receptor; ET-B; Et-b receptor; Et-b-r; ET-BR; ET-RB; Etb receptor; Etb-svr; piebald; RP11-318G21.1
Gene Aliases: ABCDS; Ednra; EDNRB; ET-B; ET-BR; ETB; ETB1; ETBR; ETR-b; ETRB; HSCR; HSCR2; Sox10m1; WS4A
UniProt ID: (Human) P24530, (Rat) P21451, (Mouse) P48302
Entrez Gene ID: (Human) 1910, (Rat) 50672, (Mouse) 13618
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support