Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LSYGSSFST SPTGQSPYTY QMHGTTGFYQ GGNGLGNAAG FGSVHQDYPS YPGFPQSQYP QYYGSSYNPP YVPASSICPS PLSTSTYVLQ EASHNVPNQS SESLAGEYNT HNGPSTPAKE GDTDRPHRAS DGKLRGRSKR SSDPSPAGDN EIERVFVWDL DETIIIFHSL LTGTFASRYG KDTTTSVRIG LMMEEMIFNL ADTHLFFNDL EDCDQIHVDD VSSDDNGQDL STYNFSADGF HSSAPGANLC LGSGVHGGVD WMRKLAFRYR RVKEMYNTYK NNVGGKESCF ERIMQRFGRK AVYVVIGDGV EEEQGAKKHN MPFWRISCHA DLEALRHALE LEYL (241-583 aa encoded by B C013882)
Functions both as protein phosphatase and as transcriptional coactivator for SIX1, and probably also for SIX2, SIX4 and SIX5. Tyrosine phosphatase that dephosphorylates 'Tyr- 142' of histone H2AX (H2AXY142ph) and promotes efficient DNA repair via the recruitment of DNA repair complexes containing MDC1. 'Tyr-142' phosphorylation of histone H2AX plays a central role in DNA repair and acts as a mark that distinguishes between apoptotic and repair responses to genotoxic stress. Its function as histone phosphatase may contribute to its function in transcription regulation during organogenesis. Plays an important role in hypaxial muscle development together with SIX1 and DACH2; in this it is functionally redundant with EYA1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Drosophila-type eyes absent 2-like protein; eyes absent 2 homolog; Eyes absent homolog 2; Protein phosphatase EYA2
Gene Aliases: EAB1; EYA2
UniProt ID: (Human) O00167, (Mouse) O08575
Entrez Gene ID: (Human) 2139, (Rat) 156826, (Mouse) 14049
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support