Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ENO1/2/3 Antibodies

          Osenses

          Enolase Polyclonal Antibody

          1 Reference
          View all (4) ENO1/2/3 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Enolase Polyclonal Antibody

          Product Details

          OSE00045W-100UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Miscellaneous PubMed (MISC)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Guinea pig / Ig

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide from aa region 200-280 of internal region of human Enolase conjugated to blue carrier protein was used as the antigen
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.

          The peptide is homologous in rat and mouse.

          Specificity of this antibody: Enolase (alpha, beta, gamma).

          Target Information

          Enolase 1 is a multifunctional enzyme that, as well as its role in glycolysis, plays a part in various processes such as growth control, hypoxia tolerance and allergic responses. May also function in the intravascular and pericellular fibrinolytic system due to its ability to serve as a receptor and activator of plasminogen on the cell surface of several cell-types such as leukocytes and neurons. Stimulates immunoglobulin production.MBP1 binds to the myc promoter and acts as a transcriptional repressor. May be a tumor suppressor.Enolase 2 has neurotrophic and neuroprotective properties on a broad spectrum of central nervous system (CNS) neurons. Binds, in a calcium-dependent manner, to cultured neocortical neurons and promotes cell survival.Enolase 3 appears to have a function in striated muscle development and regeneration.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 2-phospho-D-glycerate hydro-lyase; 2-phospho-D-glycerate hydrolase; 2-phospho-D-glycerate hydrolyase; alpha enolase; alpha enolase like 1; Alpha-enolase; Beta-enolase; c-Myc promoter binding protein; C-myc promoter-binding protein; Enolase 1; enolase 1, (alpha); enolase 1, alpha non-neuron; Enolase 2; enolase 2 (gamma, neuronal); Enolase 3; enolase 3 (beta, muscle); enolase-alpha; epididymis secretory protein Li 17; epididymis secretory protein Li 279; Gamma-enolase; MBP-1; MPB-1; MSE; muscle enriched enolase; Muscle specific enolase (beta beta enolose) (two splicing products); Muscle-specific enolase; MYC promoter-binding protein 1; Neural enolase; neuron specific gamma enolase; Neuron-specific enolase; neuronal enriched enolase; neurone-specific enolase; NNE; Non-neural enolase; NSE; Phosphopyruvate hydratase; Plasminogen-binding protein; Skeletal muscle enolase; tau-crystallin

          View more View less

          Gene Aliases: 0610008I15; AI837106; AL022784; BBE; D6Ertd375e; Eno-1; Eno-2; Eno-3; ENO1; ENO1L1; ENO2; ENO3; GSD13; HEL-S-17; HEL-S-279; MBP-1; MBPB1; MPB1; MSE; NNE; NSE; PPH; RNEN3

          View more View less

          UniProt ID: (Human) P06733, (Human) P09104, (Human) P13929, (Mouse) P17182, (Rat) P04764, (Mouse) P17183, (Rat) P07323, (Mouse) P21550, (Rat) P15429

          View more View less

          Entrez Gene ID: (Human) 2023, (Human) 2026, (Human) 2027, (Mouse) 13806, (Rat) 24333, (Mouse) 13807, (Rat) 24334, (Mouse) 13808, (Rat) 25438

          View more View less

          Function(s)
          magnesium ion binding DNA binding transcription factor activity, sequence-specific DNA binding transcription corepressor activity phosphopyruvate hydratase activity protein binding poly(A) RNA binding protein heterodimerization activity GTPase binding cadherin binding involved in cell-cell adhesion protein homodimerization activity RNA binding lyase activity metal ion binding
          Process(es)
          gluconeogenesis canonical glycolysis negative regulation of transcription from RNA polymerase II promoter in utero embryonic development transcription, DNA-templated response to virus negative regulation of cell growth negative regulation of transcription, DNA-templated positive regulation of binding cell-cell adhesion cellular response to acid chemical glycolytic process response to organic cyclic compound response to estradiol response to drug aging substantia nigra development skeletal muscle tissue regeneration
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-f7m9l:80/100.66.129.252:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0