Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • EphA2/EphA3/EphA4 Antibodies

          Invitrogen

          EphA2/EphA3/EphA4 Polyclonal Antibody

          View all (3) EphA2/EphA3/EphA4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite EphA2/EphA3/EphA4 Polyclonal Antibody

          • Antibody Testing Data (1)
          EphA2/EphA3/EphA4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          EphA2/EphA3/EphA4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          EphA2/EphA3/EphA4 Antibody (PA5-99153) in WB

          Western blot analysis of EphA2/EphA3/EphA4 in HepG2 cell lysates (left lane: treated with the antigen-specific peptide). Samples were incubated with EphA2/EphA3/EphA4 polyclonal antibody (Product # PA5-99153). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          EphA2/EphA3/EphA4 Antibody in Western Blot (WB)

          Product Details

          PA5-99153

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human EPHA2(Accession P29317), corresponding to amino acid residues R568-V618.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          RRID

          AB_2818086

          Product Specific Information

          Antibody detects endogenous levels of total EPHA2/3/4.

          Target Information

          The Eph subfamily represents the largest group of receptor protein tyrosine kinases identified to date. While the biological activities of these receptors have yet to be determined, there is increasing evidence that they are involved in central nervous system function and in development. The Eph subfamily receptors of human origin (and their murine/avian homologs) include EphA1(Eph), EphA2 (Eck), EphA3 (Hek4), EphA4 (Hek8), EphA5 (Hek7), EphA6 (Hek12),EphA7 (Hek11/MDK1), EphA8 (Hek3), EphB1 (Hek6), EphB2 (Hek5), EphB3(Cek10, Hek2), EphB4 (Htk), EphB5 (Hek9) and EphB6 (Mep). Ligands for Eph receptors include ephrin-A4 (LERK-4) which binds EphA3 and EphB1. In addition, Ephrin-A2 (Elf-1) has been described as the ligand for EphA4, ephrin-A3 (Ehk1-L) as the ligand for EphA5 and ephrin-B2 (Htk-L) as the ligand for EphB4 (Htk).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: EK4; EK8; EPH-like kinase 4; EPH-like kinase 8; Eph-like tyrosine kinase 1; Ephrin A2; Ephrin A3; Ephrin A4; ephrin receptor EphA2; Ephrin type-A receptor 2; Ephrin type-A receptor 3; Ephrin type-A receptor 4; EphrinA2; EphrinA3; EphrinA4; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; HEK; Human embryo kinase; human embryo kinase 1; receptor protein-tyrosine kinase HEK8; rEK4; soluble EPHA2 variant 1; testicular tissue protein Li 64; TYRO1 protein tyrosine kinase; TYRO4 protein tyrosine kinase; Tyrosine-protein kinase receptor ECK; Tyrosine-protein kinase receptor ETK1; tyrosine-protein kinase receptor REK4; Tyrosine-protein kinase receptor SEK; Tyrosine-protein kinase TYRO1; Tyrosine-protein kinase TYRO4

          View more View less

          Gene Aliases: ARCC2; CTPA; CTPP1; CTRCT6; ECK; EK4; EPHA2; EPHA3; EPHA4; ETK; ETK1; HEK; HEK4; HEK8; Rek4; RGD1560587; SEK; TYRO1; TYRO4

          View more View less

          UniProt ID: (Human) P29317, (Human) P29320, (Human) P54764, (Rat) O08680

          View more View less

          Entrez Gene ID: (Human) 1969, (Human) 2042, (Human) 2043, (Rat) 366492, (Rat) 29210, (Rat) 316539

          View more View less

          Function(s)
          transmembrane receptor protein tyrosine kinase activity ephrin receptor activity protein binding ATP binding cadherin binding involved in cell-cell adhesion GPI-linked ephrin receptor activity protein kinase activity transmembrane-ephrin receptor activity PH domain binding identical protein binding ephrin receptor binding DH domain binding molecular_function
          Process(es)
          cell adhesion regulation of epithelial to mesenchymal transition positive regulation of neuron projection development cell migration peptidyl-tyrosine phosphorylation regulation of actin cytoskeleton organization regulation of GTPase activity ephrin receptor signaling pathway regulation of focal adhesion assembly regulation of microtubule cytoskeleton organization cellular response to retinoic acid fasciculation of sensory neuron axon fasciculation of motor neuron axon axon guidance adult walking behavior motor neuron axon guidance glial cell migration corticospinal tract morphogenesis positive regulation of JUN kinase activity protein autophosphorylation negative regulation of axon regeneration regulation of astrocyte differentiation regulation of axonogenesis positive regulation of dendrite morphogenesis regulation of dendritic spine morphogenesis nephric duct morphogenesis positive regulation of Rho guanyl-nucleotide exchange factor activity skeletal system development angiogenesis vasculogenesis osteoblast differentiation multicellular organism development intrinsic apoptotic signaling pathway in response to DNA damage regulation of lamellipodium assembly notochord formation viral process neural tube development neuron differentiation keratinocyte differentiation osteoclast differentiation mammary gland epithelial cell proliferation regulation of cell adhesion mediated by integrin post-anal tail morphogenesis protein kinase B signaling regulation of blood vessel endothelial cell migration regulation of angiogenesis bone remodeling axial mesoderm formation negative regulation of protein kinase B signaling notochord cell development cell chemotaxis branching involved in mammary gland duct morphogenesis lens fiber cell morphogenesis regulation of ERK1 and ERK2 cascade response to growth factor positive regulation of establishment of protein localization to plasma membrane activation of GTPase activity cell-cell adhesion blood vessel development notochord morphogenesis fever generation endocardial cushion development response to lipopolysaccharide response to cytokine
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0