Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Fabgennix
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
EPHA6 is a member of the ephrin receptor subfamily of protein-tyrosine kinases which have been implicated in axon guidance, neuron-target interactions, regional compartmentalization, and synaptic functions in nervous systems. EPHA6 is highly expressed in the brain and testis. Reduction in EPHA6 has been detected in Hypospadias, a common defect affecting the growth and closure of the external genitalia. Genetic inhibition of EPHA6 in mice produces behavioral deficits specifically in tests of learning and memory. Mice deficient in EPHA6 show reduced memory of the consequences of the training context.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp434C1418; EHK-2; EK12; EPH homology kinase 2; EPH-like kinase 12; ephrin receptor EphA6; Ephrin type-A receptor 6; FLJ35246
Gene Aliases: EHK-2; EHK2; EK12; EPA6; EPHA6; HEK12; PRO57066
UniProt ID: (Human) Q9UF33
Entrez Gene ID: (Human) 285220
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support