Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive test controls include: HeLa, Mouse liver, Rat lung, Rat liver, Rat brain.
Immunogen sequence: RRHPGGSRVI SHYAGQDATD PFVAFHINKG LVKKYMNSLL IGELSPEQPS FEPTKNKELT DEFRELRATV ERMGLMKANH VFFLLYLLHI LLLDGAAWLT L
The protein encoded by this gene is a member of the fatty acid desaturase gene family. Desaturase enzymes regulate unsaturation of fatty acids through the introduction of double bonds between defined carbons of the fatty acyl chain. FADS family members are considered fusion products composed of an N-terminal cytochrome b5-like domain and a C-terminal multiple membrane-spanning desaturase portion, both of which are characterized by conserved histidine motifs. This gene is clustered with family members FADS1 and FADS2 at 11q12-q13. 1; this cluster is thought to have arisen evolutionarily from gene duplication based on its similar exon/intron organization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acyl-CoA (8-3)-desaturase; BC269730_2; D5D; delta(5) desaturase; Delta(5) fatty acid desaturase; delta-5 desaturase; delta-5 fatty acid desaturase; Fatty acid desaturase 1; FLJ38956; FLJ90273; linoleoyl-CoA desaturase (delta-6-desaturase)-like 1
Gene Aliases: 0710001O03Rik; A930006B21Rik; AI317215; D5D; DSD; FADS1; FADS6; FADSD5; LLCDL1; TU12
UniProt ID: (Human) O60427, (Mouse) Q920L1, (Rat) Q920R3
Entrez Gene ID: (Human) 3992, (Mouse) 76267, (Rat) 84575
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support