Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASNMDREMI LADFQACTGI ENIDEAITLL EQNNWDLVAA INGVIPQENG ILQSEYGGET IPGPAFNPAS HPASAPTSSS SSAFRPVMPS RQIVERQPRM LDFRVEYRDR NVDVVLEDTC TVGEIKQILE NELQIPVSKM LLKGWKTGDV EDSTVLKSLH LPKNNSLYVL TPDLPPPSSS SHAGALQESL NQNFMLIITH REVQREYNLN FSGSSTIQEV KRNVYDLTSI PVRHQLWEGW PTSATDDSMC LAESGLSYPC HRLTVGRRSS PAQTREQSEE QITDVHMVSD SDGDDFEDAT EFGVDDGEVF GMASSALRKS PMMPENAENE GDALLQFTAE FSSRYGDCHP VFFIGSLEAA FQEAFYVKAR DRKLLAIYLH HDESVLTNVF CSQMLCAESI VSYLSQNFIT WAWDLTKDSN RARFLTMCNR HFGSVVAQTI RTQKTDQFPL FLIIMGKRSS NEVLNVIQGN TTVDELMMRL MAAMEIFTAQ QQEDIKDEDE REARENVKRE QDEAYRLSLE ADRAKREAHE REMAEQFRLE QIRKEQEEER EAIRLSLEQA LPPEPKEENA EPVSKLRIRT PSGEFLERRF LASNKLQIVF DFVASKGFPW DEYKLLSTFP RRDVTQLDPN KSLLEVKLFP QETLFLEAKE
Interaction of Fas ligand (TNFSF6) with the FAS antigen (TNFRSF6) mediates programmed cell death, also called apoptosis, in a number of organ systems. The protein encoded by this gene binds to FAS antigen and can initiate apoptosis or enhance apoptosis initiated through FAS antigen. Initiation of apoptosis by the protein encoded by this gene requires a ubiquitin-like domain but not the FAS-binding domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fas (TNFRSF6) associated factor 1; FAS-associated factor 1; hFAF1; TNFRSF6-associated factor 1; UBX domain protein 3A; UBX domain-containing protein 12; UBX domain-containing protein 3A
Gene Aliases: AA408698; CGI-03; Dffrx; FAF1; Fam; hFAF1; HFAF1s; UBXD12; UBXN3A
UniProt ID: (Human) Q9UNN5, (Mouse) P54731
Entrez Gene ID: (Human) 11124, (Mouse) 14084
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support