Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTDLVAVWD VALSDGVHKI EFEHGTTSGK RVVYVDGKEE IRKEWMFKLV GKETFYVGAA KTKATINIDA ISGFAYEYTL EINGKSLKKY MEDRSKTTNT WVLHMDGENF RIVLEKDAMD VWCNGKKLET AGEFVDDGTE THFSIGNHDC YIKAVSSGKR KEGIIHTLIV DNREIPEIAS (1-179 aa encoded by BC012478)
Plays a role as an inducible effector molecule that mediates Fas resistance produced by surface Ig engagement in B cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FAIM 1; Fas apoptotic inhibitory molecule 1; rFAIM
Gene Aliases: FAIM; FAIM1
UniProt ID: (Human) Q9NVQ4, (Mouse) Q9WUD8, (Rat) Q8R5H8
Entrez Gene ID: (Human) 55179, (Mouse) 23873, (Rat) 140930
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support