Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MSVNMDELKH QVMINQFVLT AGCAADQAKQ LLQAAHWQFE TALSAFFQET NIPYSHHHHQ MMCTPANTPA TPPNFPDALT MFSRLKASES FHSGGSGSPM AATATSPPPH FPHAATSSSA ASSWPTAASP PGGPQHHQPQ PPLWTPTPPS PASDWPPLAP QQATSEPRAH PAMEAER
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 100, member A; protein FAM100A; UBA-like domain containing 1; UBA-like domain-containing protein 1
Gene Aliases: FAM100A; PP11303; UBALD1
UniProt ID: (Human) Q8TB05
Entrez Gene ID: (Human) 124402
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support