Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LPCYSSGGAP SNSGPQGHGE IHRVPTQRRP SQFDKKILLW TGRFKSMEEI PPRIPPEMID TARNKAR
Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 42%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: f162; family with sequence similarity 162, member B; Protein FAM162B
Gene Aliases: bA86F4.2; C6orf189; FAM162B
UniProt ID: (Human) Q5T6X4
Entrez Gene ID: (Human) 221303
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support