Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MEENRTKEP IQMDVEHREV LPESLPVLHN QREFSMADFL LETTVSDFGQ SHLTEEKVIS DCECVPQKHV LTSHIDEPPT QNQSDLLNKK VICKQDLECL AFTKLSETSR METFSPAVGN RIDVMGGSEE EFMKILDHLE VSCNKPSTNK TMLWNSFQIS IGISSKSFKD NDFGILHETL PEEVGHLSFN SSSSSEANFK LEPNSPMHGG TLLEDVVGGR QTTPESDFNL QALRSRYEAL KKSLSKKREE SYLSNSQTPE RHKPELSPTP QNVQTDDTLN FLDTCDLHTE HIKPSLRTSI GERKRSLSPL IKFSPVEQRL RTTIACSLGE LPNLKEEDIL NKSLDAKEPP SDLTR (602-955 aa encoded by BC010632)
HAUS6 is 1 of 8 subunits of the 390-kDa human augmin complex, or HAUS complex. The augmin complex was first identified in Drosophila, and its name comes from the Latin verb augmentare, meaning to increase. The augmin complex is a microtubule-binding complex involved in microtubule generation within the mitotic spindle and is vital to mitotic spindle assembly (Goshima et al., 2008 ; Uehara et al., 2009 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dim gamma-tubulin homolog; family with sequence similarity 29, member A; HAUS augmin-like complex subunit 6; HAUS augmin-like complex, subunit 6
Gene Aliases: DGT6; FAM29A; HAUS6; KIAA1574
UniProt ID: (Human) Q7Z4H7
Entrez Gene ID: (Human) 54801
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support