Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product # 17293-1-AP specifically recognizes the 96 kDa protein.
Immunogen sequence: KVLLLLWKV VMFTLGGFEH LQTLKVQKRA ELGLPPLAED SIQVVKSMRA ASPPSYTLDL GESQLAPPPS KLRGRRGSRR QLLTKQDSLD IYNERDLFKT EEPATEEEEE SAGDGERTLD GELDLLEQDP LVPPPPSQAP LSAERVA (268-413 aa encoded by BC019064 )
Plays a role in the regulation of cell morphology and cytoskeletal organization. Required in the control of cell shape. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 40, member B; FAR11 factor arrest 11 homolog B; homolog of yeast FAR11 protein 2; Protein FAM40B; Striatin-interacting protein 2; Striatin-interacting proteins 2
Gene Aliases: D330017J20Rik; FAM40B; FAR11B; KIAA1170; STRIP2; Stripb
UniProt ID: (Human) Q9ULQ0, (Mouse) Q8C9H6
Entrez Gene ID: (Human) 57464, (Mouse) 320609
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support