Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
FAM54A belongs to the MTFR1/FAM54 family. The function of the FAM54A protein is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DUF729 domain containing 1; DUF729 domain-containing protein 1; family with sequence similarity 54, member A; Mitochondrial fission regulator 2
Gene Aliases: DUFD1; FAM54A; MTFR2
UniProt ID: (Rat) B0K035, (Human) Q6P444
Entrez Gene ID: (Rat) 100911069, (Dog) 483997, (Human) 113115
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support