Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Required for hearing (PubMed:24958875). Involved in skeletal muscle development (PubMed:24687993).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 65, member B; myogenesis-related and NCAM-associated protein homolog; protein FAM65B; Rho family-interacting cell polarization regulator 2
Gene Aliases: Ab2-162; C6orf32; DFNB104; DIFF40; DIFF48; FAM65B; KIAA0386; MYONAP; PL48; RGD1306939; RIPOR2
UniProt ID: (Human) Q9Y4F9, (Rat) Q7TP54, (Mouse) Q80U16
Entrez Gene ID: (Human) 9750, (Rat) 306934, (Mouse) 193385
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support