Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: KFTHCLVPK MPTNSTETTP ENSLLSSPQP SEPLVLLAAE QTSGSFSQLS GKPQLTADRN NDTAIEIDNC SSYKIPSPVA SPINLNIPMR AALSHSLWEQ EDWNEHLLQV HIASYLGEHF LGA (188-309 aa encoded by BC105730)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 137, member B; family with sequence similarity 71, member F2; Golgi-associated RAB2 interactor protein 1A; Golgi-associated Rab2B interactor; protein FAM71F2
Gene Aliases: AI507611; FAM137B; FAM71F2; GARI; GARIN1A
UniProt ID: (Human) Q6NXP2, (Mouse) B2RXB0
Entrez Gene ID: (Human) 346653, (Mouse) 245884
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support