Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • FBXO2 Antibodies

          Proteintech

          FBXO2 Polyclonal Antibody

          7 References
          View all (8) FBXO2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite FBXO2 Polyclonal Antibody

          • Antibody Testing Data (24)
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 24

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FBXO2 Antibody (14590-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded mouse brain tissue slide using (Product # 14590-1-AP) (FBXO2 antibody) at dilution of 1:200 (under 10x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FBXO2 Antibody in Western Blot (WB)
          FBXO2 Antibody in Western Blot (WB)

          Product Details

          14590-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:1,000
          View 4 publications 4 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          FBXO2 Fusion Protein Ag6122 (1-296 aa encoded by BC025233)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.13 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MDGDGDPES VGQPEEASPE EQPEEASAEE ERPEDQQEEE AAAAAAYLDE LPEPLLLRVL AALPAAELVQ ACRLVCLRWK ELVDGAPLWL LKCQQEGLVP EGGVEEERDH WQQFYFLSKR RRNLLRNPCG EEDLEGWCDV EHGGDGWRVE ELPGDSGVEF THDESVKKYF ASSFEWCRKA QVIDLQAEGY WEELLDTTQP AIVVKDWYSG RSDAGCLYEL TVKLLSEHEN VLAEFSSGQV AVPQDSDGGG WMEISHTFTD YGPGVRFVRF EHGGQDSVYW KGWFGARVTN SSVWVEP (1-296 aa encoded by BC025233)

          Target Information

          This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. This protein is highly similar to the rat NFB42 (neural F Box 42 kDa) protein which is enriched in the nervous system and may play a role in maintaining neurons in a postmitotic state.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: F-box gene 1; F-box only protein 2; neural F box protein NFB42; organ of Corti protein 1; prion protein ligand 4

          View more View less

          Gene Aliases: FBG1; Fbs1; Fbs2; FBX2; FBXO2; NFB42; OCP1; Prpl4

          View more View less

          UniProt ID: (Human) Q9UK22, (Mouse) Q80UW2

          View more View less

          Entrez Gene ID: (Human) 26232, (Mouse) 230904, (Rat) 85273

          View more View less

          Function(s)
          beta-amyloid binding glycoprotein binding ubiquitin-protein transferase activity protein binding carbohydrate binding
          Process(es)
          protein polyubiquitination cellular protein modification process proteolysis glycoprotein catabolic process negative regulation of cell proliferation protein ubiquitination ER-associated ubiquitin-dependent protein catabolic process SCF-dependent proteasomal ubiquitin-dependent protein catabolic process regulation of protein ubiquitination ubiquitin-dependent protein catabolic process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-2w6sl:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0