Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MYLPPHDPHV DDPMRFKPLF RIHLMERKAA TVECMYGHKG PHHGHIQIVK KDEFSTKCNQ TDHHRMSGGR QEEFRTWLRE EWGRTLEDIF HEHMQELILM KFIYTSQYDN CLTYRRIYLP PSRPDDLIKP GLFKGTYGSH GLEIVMLSFH GRRARGTKIT GDPNIPAGQQ TVEIDLRHRI QLPDLENQRN FNELSRIVLE VRERVRQEQQ EGGHEAGEGR GRQGPRESQP SPAQPRAEAP SKGPDGTPGE DGGEPGDAVA AAEQPAQCGQ GQPFVLPVGV SSRNEDYPRT CRMCFYGTGL IAGHGFTSPE RTPGVFILFD EDRFGFVWLE LKSFSLYSRV QATFRNADAP SPQAFDEMLK NIQSLTS
Members of the F-box protein family, such as FBXO31, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, cullin, and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp434B027; DKFZp434J1815; F-box only protein 31; FLJ22477; MGC15419; MGC9527; putative breast cancer tumor-suppressor; SCF ubiquitin ligase specificity factor
Gene Aliases: FBX14; FBX31; FBXO14; FBXO31; MRT45; PP2386
UniProt ID: (Human) Q5XUX0
Entrez Gene ID: (Human) 79791
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support