Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MPFLGQDWRS PGQNWVKTAD GWKRFLDEKS GSFVSDLSSY CNKEVYNKEN LFNSLNYDVA AKKRKKDMLN SKTKTQYFHQ EKWIYVHKGS TKERHGYCTL GEAFNRLDFS TAILDSRRFN YVVRLLELIA KSQLTSLSGI AQKNFMNILE KVVLKVLEDQ QNIRLIRELL QTLYTSLCTL VQRVGKSVLV GNINMWVYRM ETILHWQQQL NNIQITRPAF KGLTFTDLPL CLQLNIMQRL SDGRDLVSLG QAAPDLHVLS EDRLLWKKLC QYHFSERQIR KRLILSDKGQ LDWKKMYFKL VRCYPRKEQY GDTLQLCKHC HILSWKGTDH PCTANNPESC SVSLSPQDFI NLFKF
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs, which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and contains an F-box domain. This protein is highly expressed during muscle atrophy, whereas mice deficient in this gene were found to be resistant to atrophy. This protein is thus a potential drug target for the treatment of muscle atrophy. Alternative splicing of this gene results in two transcript variants encoding two isoforms of different sizes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: atrogin 1; Atrogin-1; F-box only protein 32; fbxo25; FLJ32424; MAFbx; MGC108443; MGC137646; MGC33610; Muscle atrophy F-box protein
Gene Aliases: Fbx32; FBXO32; MAFbx
UniProt ID: (Human) Q969P5
Entrez Gene ID: (Human) 114907
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support