Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: VCFRNYNAK TENLAPVKNQ LQIQEEEETL QDEEVWDALT DNYIPSLSED WRDPNIEALN GNCSDTEIHE KEEEEFNEKS ENDSGINEEP LLTADQVIEE IEEMMQNSPD PEEEEEVLEE EDGGETSSQA DSVLLQEMQA LTQTFNNNWS YEGLRHMSGS ELTELLDQVE GAIRDFSEEL VQQLARRDEL EFEKEVKNSF ITVLIEVQNK QKEQRELMKK RRKEKGLSLQ SSRIEKGNQM PLKRFSMEGI SNILQSGIRQ TFGSSGTDKQ YLNTVIPYEK KASPPSVEDL QMLTNILFAM KEDNEKVPTL LTDYILKVLC PT (72-392 aa encoded by BC009545)
This gene is an ortholog of the C. elegans unc-76 gene, which is necessary for normal axonal bundling and elongation within axon bundles. Expression of this gene in C. elegans unc-76 mutants can restore to the mutants partial locomotion and axonal fasciculation, suggesting that it also functions in axonal outgrowth. The N-terminal half of the gene product is highly acidic. Alternatively spliced transcript variants encoding different isoforms of this protein have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: F37; fasciculation and elongation protein zeta 1 (zygin I); Fasciculation and elongation protein zeta-1; protein kinase C-binding protein Zeta1; zygin 1; Zygin I; Zygin-1
Gene Aliases: FEZ1; UNC-76; UNC76
UniProt ID: (Human) Q99689, (Rat) P97577, (Mouse) Q8K0X8
Entrez Gene ID: (Human) 9638, (Rat) 81730, (Mouse) 235180
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support