Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 92%; Horse: 85%; Human: 100%; Mouse: 86%; Pig: 85%; Rabbit: 79%; Rat: 100%
FGD3 gene ontology annotations related to this gene include regulation of Rho protein signal transduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: faciogenital dysplasia 3; FGD1 family, member 3; FLJ00004; FYVE, RhoGEF and PH domain-containing protein 3; Zinc finger FYVE domain-containing protein 5
Gene Aliases: FGD3; ZFYVE5
UniProt ID: (Human) Q5JSP0
Entrez Gene ID: (Human) 89846
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support