Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
The antibody detects a 140 kDa* protein corresponding to the apparent molecular mass of FHOD1 on SDS-PAGE immunoblots of human K562 and Jurkat, and has weak reactivity to FHOD1 in mouse C2C12.
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FH1/FH2 domain-containing protein 1; FHOS; Formin homolog overexpressed in spleen 1; Formin homology 2 domain-containing protein 1
Gene Aliases: FHOD1; FHOS; FHOS1
UniProt ID: (Human) Q9Y613
Entrez Gene ID: (Human) 29109
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support