Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • FITC/5-FAM/6-FAM Antibodies

          Invitrogen

          FITC/5-FAM/6-FAM Recombinant Rabbit Monoclonal Antibody (4L8R5)

          View all (2) FITC/5-FAM/6-FAM antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite FITC/5-FAM/6-FAM Recombinant Rabbit Monoclonal Antibody (4L8R5)

          • Antibody Testing Data (2)
          FITC/5-FAM/6-FAM Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FITC/5-FAM/6-FAM Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FITC/5-FAM/6-FAM Antibody (MA5-55271) in WB

          FITC/5-FAM/6-FAM Recombinant Monoclonal Antibody (Product # MA5-55271) was tested in Western blot against FITC Goat IgG, TRITC Goat IgG, and Goat IgG at 1:4,000 dilution followed by a HRP-conjugated Goat anti-Rabbit IgG (H+L) secondary antibody at 1:10,000 dilution. Lysates/proteins: 25 μg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 10s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FITC/5-FAM/6-FAM Antibody in Western Blot (WB)
          FITC/5-FAM/6-FAM Antibody in ELISA (ELISA)

          Product Details

          MA5-55271

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000-1:4,800
          -

          ELISA (ELISA)

          1:500-1:2,000
          -
          Product Specifications

          Species Reactivity

          Chemical

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          4L8R5

          Immunogen

          FITC

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.05% BSA

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          RRID

          AB_3667513

          Product Specific Information

          Immunogen Sequence: FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju FITC conju

          Target Information

          FITC (Fluorescein Isothiocyanate), 5-FAM (5-Carboxyfluorescein), and 6-FAM (6-Carboxyfluorescein) are commonly used gene targets for fluorescent labeling in molecular biology research. These dyes belong to the fluorescein family and emit a green fluorescence when excited by specific wavelengths of light. FITC, 5-FAM, and 6-FAM are utilized in various techniques such as fluorescence in situ hybridization (FISH), flow cytometry, immunofluorescence, and DNA sequencing. By attaching these dyes to gene-specific probes or primers, researchers can visualize and detect gene expression levels, identify specific DNA or RNA sequences, and study cellular processes. The compatibility of FITC, 5-FAM, and 6-FAM with different imaging systems and their bright green fluorescence make them valuable tools in molecular biology, aiding in the understanding of gene function and cellular dynamics.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline