Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PPIases accelerate the folding of proteins during protein synthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 19 kDa FK506-binding protein; 19 kDa FKBP; FK506 binding protein 11, 19 kDa; FK506-binding protein 11; FKBP-11; FKBP-19; Peptidyl-prolyl cis-trans isomerase FKBP11; PPIase FKBP11; Rotamase
Gene Aliases: 1110002O23Rik; FKBP-11; FKBP11; FKBP19; UNQ336/PRO535
UniProt ID: (Human) Q9NYL4, (Mouse) Q9D1M7
Entrez Gene ID: (Human) 51303, (Mouse) 66120, (Rat) 300211
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support