Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTLRPSLLPL HLLLLLLLSA AVCRAEAGLE TESPVRTLQV ETLVEPPEPC AEPAAFGDTL HIHYTGSLVD GRIIDTSLTR DPLVIELGQK QVIPGLEQSL LDMCVGEKRR AIIPSHLAYG KRGFPPSVPA DAVVQYDVEL IALIRANYWL KLVKGILPLV GMAMVPALLG LIGYHLYRKA NRPKVSKKKL KEEKRNKSKK K
PPIases accelerate the folding of proteins during protein synthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 19 kDa FK506-binding protein; 19 kDa FKBP; FK506 binding protein 11, 19 kDa; FK506-binding protein 11; FKBP-11; FKBP-19; Peptidyl-prolyl cis-trans isomerase FKBP11; PPIase FKBP11; Rotamase
Gene Aliases: FKBP11; FKBP19; UNQ336/PRO535
UniProt ID: (Human) Q9NYL4
Entrez Gene ID: (Human) 51303
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support