Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDTETTALCP SGSRRASPPG TPTPEADATL LKKSEKLLAE LDRSGLPSAP GAPRRRGSMP VPYKHQLRRA QAVDELDWPP QASSSGSSDS LGSGEAAPAQ KDGIFKVMLV GESGVGKSTL AGTFGGLQGD SAHEPENPED TYERRIMVDK EEVTLVVYDI WEQGDAGGWL RDHCLQTGDA FLIVFSVTDR RSFSKVPETL LRLRAGRPHH DLPVILVGNK SDLARSREVS LEEGRHLAGT LSCKHIETSA ALHHNTRELF EGAVRQIRLR RGRNHAGGQR PDPGSPEGPA PPARRESLTK KAKRFLANLV PRNAKFFKQR SRSCHDLSVL
Binds GTP saturably and exhibits a low intrinsic rate of GTP hydrolysis. Target protein is a Ras-related small GTP-binding protein that is found in high levels in neuronal tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP-binding protein REM 2; Rad and Gem-like GTP-binding protein 2; Rad and Gem-related 2 (rat homolog); RAS (RAD and GEM)-like GTP binding 2; RAS-like GTP binding 2
Gene Aliases: REM2
UniProt ID: (Human) Q8IYK8
Entrez Gene ID: (Human) 161253
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support