Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Filamin A Antibodies

          Abnova

          FLNA Monoclonal Antibody (4E10-1B2)

          View all (28) Filamin A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite FLNA Monoclonal Antibody (4E10-1B2)

          • Antibody Testing Data (4)
          FLNA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FLNA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FLNA Antibody (H00002316-M01) in IHC (P)

          Immunoperoxidase of monoclonal antibody to FLNA on formalin-fixed paraffin-embedded human lymphoma tissue. Antibody concentration is 1.5 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FLNA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FLNA Antibody in Western Blot (WB)
          FLNA Antibody in ELISA (ELISA)
          FLNA Antibody in Proximity Ligation Assay (PLA) (PLA)

          Product Details

          H00002316-M01

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1.5 µg/mL
          -

          ELISA (ELISA)

          0.03 ng/mL
          -

          in situ PLA (PLA)

          1:1200
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          4E10-1B2

          Immunogen

          FLNA (AAH14654, 1 a.a. approximately 838 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MPSGKVAQPT ITDNKDGTVT VRYAPSEAGL HEMDIRYDNM HIPGSPLQFY VDYVNCGHVT AYGPGLTHGV VNKPATFTVN TKDAGEGGLS LAIEGPSKAE ISCTDNQDGT CSVSYLPVLP GDYSILVKYN EQHVPGSPFT ARVTGDDSMR MSHLKVGSAA DIPINISETD LSLLTATVVP PSGREEPCLL KRLRNGHVGI SFVPKETGEH LVHVKKNGQH VASSPIPVVI SQSEIGDASR VRVSGQGLHE GHTFEPAEFI IDTRDAGYGG LSLSIEGPSK VDINTEDLED GTCRVTYCPT EPGNYIINIK FADQHVPGSP FSVKVTGEGR VKESITRRRR APSVANVGSH CDLSLKIPEI SIQDMTAQVT SPSGKTHEAE IVEGENHTYC IRFVPAEMGT HTVSVKYKGQ HVPGSPFQFT VGPLGEGGAH KVRAGGPGLE RAEAGVPAEF SIWTREAGAG GLAIAVEGPS KAEISFEDRK DGSCGVAYVV QEPGDYEVSV KFNEEHIPDS PFVVPVASPS GDARRLTVSS LQESGLKVNQ PASFAVSLNG AKGAIDAKVH SPSGALEECY VTEIDQDKYA VRFIPRENGV YLIDVKFNGT HIPGSPFKIR VGEPGHGGDP GLVSAYGAGL EGGVTGNPAE FVVNTSNAGA GALSVTIDGP SKVKMDCQEC PEGYRVTYTP MAPGSYLISI KYGGPYHIGG SPFKAKVTGP RLVSNHSLHE TSSVFVDSLT KATCAPQHGA PGPGPADASK VVAKGLGLSK AYVGQKSSFT VDCSKAGNNM LLVGVHGPRT PCEEILVKHV GSRLYSVSYL LKDKGEYTLV VKWGDEHIPG SPYRVVVP

          Target Information

          Filamin functions as a crosslinking protein forming a flexible link between two actin filaments. It is composed of two identical polypeptide chains each joined to the other at one end, with an actin binding site at the other. It is present in human platelets, lymphocytes, fibroblasts and smooth muscle actin.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ABP-280; actin binding protein 280; Actin-binding protein 280; alpha filamin; Alpha-filamin; Endothelial actin-binding protein; filamin 1; filamin A, alpha; Filamin-1; Filamin-A; Filamin1; FilaminA; FLN-A; Non-muscle filamin

          View more View less

          Gene Aliases: ABP-280; ABPX; CSBS; CVD1; FLN; FLN-A; FLN1; FLNA; FMD; MNS; NHBP; OPD; OPD1; OPD2; XLVD; XMVD

          View more View less

          UniProt ID: (Human) P21333

          View more View less

          Entrez Gene ID: (Human) 2316

          View more View less

          Function(s)
          G-protein coupled receptor binding glycoprotein binding signal transducer activity protein binding transcription factor binding Rho GTPase binding Ral GTPase binding kinase binding small GTPase binding mu-type opioid receptor binding Fc-gamma receptor I complex binding protein homodimerization activity poly(A) RNA binding SMAD binding Rac GTPase binding actin filament binding GTPase binding cadherin binding involved in cell-cell adhesion
          Process(es)
          platelet degranulation adenylate cyclase-inhibiting dopamine receptor signaling pathway negative regulation of transcription from RNA polymerase I promoter platelet activation regulation of cell migration actin cytoskeleton reorganization cell junction assembly protein localization to cell surface negative regulation of protein catabolic process cilium assembly mRNA transcription from RNA polymerase II promoter positive regulation of transcription factor import into nucleus negative regulation of apoptotic process receptor clustering positive regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of sequence-specific DNA binding transcription factor activity wound healing, spreading of cells establishment of protein localization protein stabilization cytoplasmic sequestering of protein actin crosslink formation platelet aggregation semaphorin-plexin signaling pathway mitotic spindle assembly positive regulation of substrate adhesion-dependent cell spreading positive regulation of integrin-mediated signaling pathway
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0