Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPGSFVPTVT AITTSQDLQW LVQPTLISSM AQSQGQPLAS QPPVVDPYDM PGTSYSTPGM SGYSSGGASG SGGPSTSGTT SGPGPARPAR ARPRRPREET LTPEEEEKRR VRRERNKLAA AKCRNRRREL TDRLQAETDQ LEEEKAELES EIAELQKEKE RLEFVLVAHK PGCKIPYEEG PGPGPLAEVR DLPGSAPAKE DGFSWLLPPP PPPPLPFQTS QDAPPNLTAS LFTHSEVQVL GDPFPVVNPS YTSSFVLTCP EVSAFAGAQR TSGSDQPSDP LNSPSLLAL
c-FOS is a nuclear phosphoprotein which forms a tight but non-covalently linked complex with the JUN/AP-1 transcription factor. It has a critical function in regulating the development of cells destined to form and maintain the skeleton, and is thought to have an important role in signal transduction, cell proliferation and differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activator protein 1; DKFZp686C0818; FBJ murine osteosarcoma viral oncogene homolog B; fos b; FosB proto-oncogene, AP-1 trancription factor subunit; FosB proto-oncogene, AP-1 transcription factor subunit; G0/G1 switch regulatory protein 3; MGC42291; oncogene FOS-B; Protein FosB; Transcription factor AP-1 subunit FosB
Gene Aliases: AP-1; FOSB; G0S3; GOS3; GOSB
UniProt ID: (Human) P53539
Entrez Gene ID: (Human) 2354
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support