Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRSRPAGPAL LLLLLFLGAA ESVRRAQPPR RYTPDWPSLD SRPLPAWFDE AKFGVFIHWG VFSVPAWGSE WFWWHWQGEG RPQYQRFMRD NYPPGFSYAD FGPQFTARFF HPEEWADLFQ AAGAKYVVLT TKHHEGFTNW PSPVSWNWNS KDVGPHRDLV GELGTALRKR NIRYGLYHSL LEWFHPLYLL DKKNGFKTQH FVSAKTMPEL YDLVNSYKPD LIWSDGEWEC PDTYWNSTNF LSWLYNDSPV KDEVVVNDRW GQNCSCHHGG YYNCEDKFKP QSLPDHKWEM CTSIDKFSWG YRRDMALSDV TEESEIISEL VQTVSLGGNY LLNIGPTKDG LIVPIFQERL LAVGKWLSIN GEAIYASKPW RVQWEKNTTS VWYTSKGSAV YAIFLHWPEN GVLNLESPIT TSTTKITMLG IQGDLKWSTD PDKGLFISLP QLPPSAVPAE FAWTIKLTGV K
Alpha-L-fucosidase (EC 3.2.1.51) is a lysosomal enzyme involved in the degradation of fucose-containing glycoproteins and glycolipids (Occhiodoro et al., 1989 [PubMed 2803312]). At least 2 separate polymorphic alpha-L-fucosidases are recognized in man: that in tissues, FUCA1, which is deficient in fucosidosis (MIM 230000), and that in plasma, FUCA2 (MIM 136820).[supplied by OMIM].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A L fucosidase I; A L fucoside fucohydrolase 1; Alpha-L-fucosidase 1; Alpha-L-fucosidase I; Alpha-L-fucoside fucohydrolase 1; EC 3.2.1; EC 3.2.1.51; Tissue alpha-L-fucosidase; Tissue α L fucosidase; α L fucosidase 1; α L fucosidase I; α L fucoside fucohydrolase 1
Gene Aliases: FUCA; FUCA1; Nbla10230
UniProt ID: (Human) P04066
Entrez Gene ID: (Human) 2517
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support