Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Alpha-L-fucosidase (EC 3.2.1.51) is a lysosomal enzyme involved in the degradation of fucose-containing glycoproteins and glycolipids (Occhiodoro et al., 1989 [PubMed 2803312]). At least 2 separate polymorphic alpha-L-fucosidases are recognized in man: that in tissues, FUCA1, which is deficient in fucosidosis (MIM 230000), and that in plasma, FUCA2 (MIM 136820).[supplied by OMIM].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A L fucosidase I; A L fucoside fucohydrolase 1; Alpha-L-fucosidase 1; Alpha-L-fucosidase I; Alpha-L-fucoside fucohydrolase 1; EC 3.2.1; EC 3.2.1.51; fucosidase, alpha-L-1, tissue; Tissue alpha-L-fucosidase; Tissue α L fucosidase; α L fucosidase 1; α L fucosidase I; α L fucoside fucohydrolase 1
Gene Aliases: 0610006A03Rik; 9530055J05Rik; Afuc; FUCA; FUCA1; Nbla10230
UniProt ID: (Human) P04066, (Mouse) Q99LJ1
Entrez Gene ID: (Human) 2517, (Mouse) 71665
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support