Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • FXR2 Antibodies

          Proteintech

          FXR2 Polyclonal Antibody

          2 References
          View all (7) FXR2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite FXR2 Polyclonal Antibody

          • Antibody Testing Data (4)
          FXR2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FXR2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FXR2 Antibody (12552-1-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells using 12552-1-AP (FXR2 antibody) at dilution of 1:25 and Rhodamine-Goat anti-Rabbit IGG. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FXR2 Antibody in Immunocytochemistry (ICC/IF)
          FXR2 Antibody in Western Blot (WB)
          FXR2 Antibody in Western Blot (WB)
          FXR2 Antibody in Immunoprecipitation (IP)

          Product Details

          12552-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -

          RNA Immunoprecipitation (RIP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          FXR2 Fusion Protein Ag3402 (1-354 aa encoded by BC020090)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.27 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MGGLASGGD VEPGLPVEVR GSNGAFYKGF VKDVHEDSVT IFFENNWQSE RQIPFGDVRL PPPADYNKEI TEGDEVEVYS RANEQEPCGW WLARVRMMKG DFYVIEYAAC DATYNEIVTL ERLRPVNPNP LATKGSFFKV TMAVPEDLRE ACSNENVHKE FKKALGANCI FLNITNSELF ILSTTEAPVK RASLLGDMHF RSLRTKLLLM SRNEEATKHL ETSKQLAAAF QEEFTVREDL MGLAIGTHGA NIQQARKVPG VTAIELGEET CTFRIYGETP EACRQARSYL EFSEDSVQVP RNLVGKVIGK NGKVIQEIVD KSGVVRVRVE GDNDKKNPRE EGMVPFIFVG TRENI (1-354 aa encoded by BC020090)

          Target Information

          Fragile X syndrome is caused by the absence of the fragile X mental-retardation protein (FMRP). FMRP is the archetype of a class of cytoplasmic mRNA-binding proteins that includes the fragile X-related 1 and 2 proteins (FXR1 and FXR2). The fragile X-related proteins FXR1 and FXR2 contain a functional nucleolar-targeting signal equivalent to the HIV-1 regulatory proteins.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: FMR1 autosomal homolog 2; fragile X mental retardation gene 2, autosomal homolog; fragile X mental retardation syndrome-related protein 2; fragile X mental retardation, autosomal homolog 2; fragile X mental retardation-related protein 2; fragile X-mental retardation 1-like 2; FXR2P; RNA-binding protein FXR2

          View more View less

          Gene Aliases: FMR1L2; FXR2; Fxr2h; FXR2P

          View more View less

          UniProt ID: (Human) P51116

          View more View less

          Entrez Gene ID: (Human) 9513, (Mouse) 23879

          View more View less

          Function(s)
          RNA binding mRNA binding protein binding identical protein binding protein homodimerization activity poly(A) RNA binding protein heterodimerization activity nucleic acid binding
          Process(es)
          positive regulation of defense response to virus by host negative regulation of translation xenophagy
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0