Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: CHEEQNSVVC SCARGYTLAD NGKACIPTGP YPCGKQTLER RKRSVAQATS SSGEAPDSIT WKPYDAADLD PTENPFDLLD FNQTQPERGD NNLTRIVGGQ ECKDGECPWQ ALLINEENEG FCGGTILSEF YILTAAHCLY QAKRFKVRVG DRNTEQEEGG EAVHEVEVVI KHNRFTKETY DFDIAVLRLK TPITFRMNVA PACLPERDWA ESTLMTQKTG IVSGFGRTHE KGRQSTRLKM LEVPYVDRNS CKLSSSFIIT QNMFCAGYDT KQEDACQGDS GGPHVTRFKD TYFVTGIVSW GEGCARKGKY GIYTKVTAFL KWIDRSMKTR GLPKAKSHAP EVITSSPLK
F10 undergoes multiple processing steps before its preproprotein is converted to a mature two-chain form by the excision of the tripeptide RKR. Two chains of the factor are held together by 1 or more disulfide bonds; the light chain contains 2 EGF-like domains, while the heavy chain contains the catalytic domain which is structurally homologous to those of the other hemostatic serine proteases. The mature factor is activated by the cleavage of the activation peptide by factor IXa (in the intrisic pathway), or by factor VIIa (in the extrinsic pathway). The activated factor then converts prothrombin to thrombin in the presence of factor Va, Ca+2, and phospholipid during blood clotting. Mutations of this gene result in factor X deficiency, a hemorrhagic condition of variable severity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coagulation factor X; Factor 10; factor Xa; Factor10; prothrombinase; RP11-98F14.9; Stuart factor; Stuart-Prower factor
Gene Aliases: F10; FX; FXA
UniProt ID: (Human) P00742
Entrez Gene ID: (Human) 2159
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support