Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive test controls include: Mouse brain, Mouse thymus, Rat heart. The target is usually found in the following locations: Cytoplasm, Nucleus.
Immunogen sequence: RKLPVFKSLR HMRQVLGAPS FRMLAWHVLM GNQVIWKSRD VDLVQSAFEV LRTMLPVGCV RIIPYSSQYE EAYRCNFLGL SPHVQIPPHV LSSEFAVIVE VHAAARSTLH PVGCEDDQSL SKYEFVVTSG SPVAADRVGP TILNKIEAAL TNQNLSVDVV DQCLVCLKEE WMNKVKVLFK FTKVDSRPKE DTQKLLSILG ASEEDNVKLL KFWMTGLSKT YKSHLMSTVR SPTASESRN
This gene encodes a mitochondrial protein that is characterized by a poly-proline rich region. A chicken homolog of this protein promotes mitochondrial fission and the mouse homolog protects cells from oxidative stress. A related pseudogene of this gene is found on chromosome X.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BHD skin lesion fibrofolliculoma protein; Birt-Hogg-Dube syndrome protein; Birt-Hogg-Dube syndrome protein homolog; Folliculin
Gene Aliases: AU014660; B430214A04Rik; BHD; FLCL; FLCN
UniProt ID: (Human) Q8NFG4, (Mouse) Q8QZS3, (Rat) Q76JQ2
Entrez Gene ID: (Human) 201163, (Mouse) 216805, (Rat) 303185
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support