Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: KNGAGLSKS KGSRIGFDST QWRAVKKFIM LTSNQNVPVF LIDPLILELI NKNFEQVKNT SHGSTSQCKF FCVPRDFTAF ALQYHLWKNE EGWFRIAENM GFQCLKIESK DPRLDGIDSL SGTEIPLHYI CKLATHAIHL VVFHERSGNY LWHGHLRLKE HIDRKFVPFR KLQFGRYPGA FDRPELQQVT VDGLEVLIPK DPMHFVEEVP HSRFIECRYK EARAFFQQYL DDNTVEAVAF RKSAKELLQL AAKTLNKLGV PFWLSSGTCL GWYRQCNIIP YSKDVDLGIF IQDYKSDIIL AFQDAGLPLK HKFGKVEDSL ELSFQGKDDV KLDVFFFYEE TDHMWNGGTQ AKTGKKFKYL FPKFTLCWTE FVDMKVHVPC ETLEYIEANY GKTWKIPVKT WDWKRSPPNV QPNGIWPISE WDEVIQLY (35-461 aa encoded by BC101808 )
FKTN regulates the migration and assembly of neurons during cortical histogenesis. Fukuyama congenital muscular dystrophy results from mutations in its gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fukutin; Fukuyama type congenital muscular dystrophy (fukutin); Fukuyama type congenital muscular dystrophy homolog; Fukuyama type congenital muscular dystrophy protein; Fukuyama-type congenital muscular dystrophy protein; patient fukutin; Ribitol-5-phosphate transferase; Ribitol-5-phosphate transferase FKTN; RP11-235C23.1
Gene Aliases: CMD1X; D830030O17Rik; FCMD; FKTN; LGMD2M; MDDGA4; MDDGB4; MDDGC4
UniProt ID: (Human) O75072, (Mouse) Q8R507
Entrez Gene ID: (Human) 2218, (Rat) 362520, (Mouse) 246179
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support