Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MSGGGDVVCT GWLRKSPPEK KLRRYAWKKR WFILRSGRMS GDPDVLEYYK NDHSKKPLRI INLNFCEQVD AGLTFNKKEL QDSFVFDIKT SERTFYLVAE TEEDMNKWVQ SICQICGFNQ AEESTDSLRN VSSAGHGPRS SPAELSSSSQ HLLRERKSSA PSHSSQPTLF TFEPPVSNHM QPTLSTSAPQ EYLYLHQCIS RRAENARSAS FSQGTRASFL MRSDTAVQKL AQGNGHCVNG ISGQVHGFYS LPKPSRHNTE FRDSTYDLPR SLASHGHTKG SLTGSETDNE DVYTFKTPSN TLCREFGDLL VDNMDVPATP LSAYQIPRTF TLDKNHNAMT VATPGDSAIA PPPRPPKPSQ AETPRWGSPQ QRPPISENSR SVAATIPRRN TLPAMDNSRL HRASSCETYE YPQRGGESAG RSAESMSDGV GSFLPGKMIV GRSDSTNSED NYVPMNPGSS TLLAMERAGD NSQSVYIPMS PGAHHFDSLG YPSTTLPVHR GPSRGSEIQP PPVNRNLKPD RKAKPTPLDL RNNTVIDELP FKSPITKSWS RANHTFNSSS SQYCRPISTQ SITSTDSGDS EENYVPMQNP VSASPVPSGT NSPAPKKSTG SVDYLALDFQ PSSPSPHRKP STSSVTSDEK VDYVQVDKEK TQALQNTMQE WTDVRQSSEP SKGAKL
This gene is a member of the GRB2-associated binding protein (GAB) gene family. These proteins contain pleckstrin homology (PH) domain, and bind SHP2 tyrosine phosphatase and GRB2 adapter protein. They act as adapters for transmitting various signals in response to stimuli through cytokine and growth factor receptors, and T- and B-cell antigen receptors. The protein encoded by this gene is the principal activator of phosphatidylinositol-3 kinase in response to activation of the high affinity IgE receptor. Two alternatively spliced transcripts encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GRB2-associated binder 2; GRB2-associated binding protein 2; GRB2-associated-binding protein 2; GRB3-3; Growth factor receptor bound protein 2-associated protein 2; pp100
Gene Aliases: GAB2; KIAA0571
UniProt ID: (Human) Q9UQC2
Entrez Gene ID: (Human) 9846
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support