Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Additional Information: This antibody is expected to recognize reported isoforms a, b and c (NP_001461.1, NP_068703.1, NP_068704.2 resp.).
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:4,000.
Receptor for GABA. The activity of this receptor is mediated by G-proteins that inhibit adenylyl cyclase activity, stimulates phospholipase A2, activates potassium channels, inactivates voltage-dependent calcium-channels and modulates inositol phospholipids hydrolysis. Plays a critical role in the fine-tuning of inhibitory synaptic transmission. Pre-synaptic GABA-B-R inhibit neurotransmitter release by down-regulating high-voltage activated calcium channels, whereas postsynaptic GABA-B-R decrease neuronal excitability by activating a prominent inwardly rectifying potassium (Kir) conductance that underlies the late inhibitory postsynaptic potentials. Not only implicated in synaptic inhibition but also in hippocampal long-term potentiation, slow wave sleep, muscle relaxation and antinociception.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAAP-188P13.3; dJ271M21.1.1; dJ271M21.1.2; FLJ92613; GABA(B) receptor; GABA-B receptor 1; GABA-B receptor, R1 subunit; Gaba-b-r1a; Gaba-b1; Gaba-br1a receptor; GABAB; Gababr1 receptor; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) B receptor, 1; Gamma-aminobutyric acid type B receptor subunit 1; GBR1; HGB1a; OTTHUMP00000109099; OTTHUMP00000214725; seven transmembrane helix receptor
Gene Aliases: GABABR1; GABBR1; GABBR1-3; GB1; GPRC3A
UniProt ID: (Human) Q9UBS5
Entrez Gene ID: (Human) 2550
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support