Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
GABRG3 is a family of proteins involved in the GABAergic neurotransmission of the mammalian central nervous system. GABRG3 is a member of the GABA-A receptor gene family of heteromeric pentameric ligand-gated ion channels through which GABA, the major inhibitory neurotransmitter in the mammalian brain, acts. GABA-A receptors are the site of action of a number of important pharmacologic agents including barbiturates, benzodiazepines, and ethanol (summary by Whiting et al., 1999). For additional general information about the GABA-A receptor gene family, see GABRA1 (MIM 137160).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAE2; ECA2; GABA(A) receptor subunit gamma-3; GABA(G) receptor; GABA(G) receptor, gamma 3; GABA-alpha receptor gamma-3 subunit; GABAAR subunit gamma-3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, gamma 3; gamma-aminobutyric acid (GABA) A receptor, subunit gamma 3; gamma-aminobutyric acid (GABA-A) receptor, subunit gamma 3; gamma-aminobutyric acid A receptor, gamma 3; Gamma-aminobutyric acid receptor subunit gamma-3; GEFSP3
Gene Aliases: B230362M20Rik; Gabrg-3; GABRG3
UniProt ID: (Human) Q99928, (Rat) P28473, (Mouse) P27681
Entrez Gene ID: (Human) 2567, (Rat) 79211, (Mouse) 14407
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support