Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRRSVYCKVV LATSLMWVLV DVFLLLYFSE CNKCDDKKER SLLPALRAVI SRNQEGPGEM GKAVLIPKDD QEKMKELFKI NQFNLMASDL IALNRSLPDV RLEGCKTKVY PDELPNTSVV IVFHNEAWST LLRTVYSVIN RSPHYLLSEV ILVDDASERD FLKLTLENYV KNLEVPVKII RMEERSGLIR ARLRGAAASK GQVITFLDAH CECTLGWLEP LLARIKEDRK TVVCPIIDVI SDDTFEYMAG SDMTYGGFNW KLNFRWYPVP QREMDRRKGD RTLPVRTPTM AGGLFSIDRN YFEEIGTYDA GMDIWGGENL EMSFRIWQCG GSLEIVTCSH VGHVFRKATP YTFPGGTGHV INKNNRRLAE VWMDEFKDFF YIISPGVVKV DYGDVSVRKT LRENLKCKPF SWYLENIYPD SQIPRRYYSL GEIRNVETNQ CLDNMGRKEN EKVGIFNCHG MGGNQVFSYT ADKEIRTDDL CLDVSRLNGP VIMLKCHHMR GNQLWEYDAE RLTLRHVNSN QCLDEPSEED KMVPTMQDCS GSRSQQWLLR NMTLGT
The GALNT13 protein is a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAcT; EC 2.4.1.41) family, which initiate O-linked glycosylation of mucins by the initial transfer of N-acetylgalactosamine (GalNAc) with an alpha-linkage to a serine or threonine residue.The GALNT13 protein is a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAcT; EC 2.4.1.41) family, which initiate O-linked glycosylation of mucins (see MUC3A, MIM 158371) by the initial transfer of N-acetylgalactosamine (GalNAc) with an alpha-linkage to a serine or threonine residue.[supplied by OMIM].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ16031; FLJ41157; GalNAc transferase 13; GalNAc-T13; H_NH0187G20.1; MGC119459; MGC119461; Polypeptide GalNAc transferase 13; Polypeptide N-acetylgalactosaminyltransferase 13; pp-GaNTase 13; Protein-UDP acetylgalactosaminyltransferase 13; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 13; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 13 (GalNAc-T13); WUGSC:H_NH0187G20.1
Gene Aliases: GalNAc-T13; GALNT13; KIAA1918
UniProt ID: (Human) Q9UIE5
Entrez Gene ID: (Human) 114805
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support