Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody detects endogenous levels of total GANP / MCM3AP protein.
POLE3 is a histone-fold protein that interacts with other histone-fold proteins to bind DNA in a sequence-independent manner. These histone-fold protein dimers combine within larger enzymatic complexes for DNA transcription, replication, and packaging.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 80 kDa MCM3-associated protein; FLJ44336; FLJ45306; GANP; germinal center-associated nuclear protein; Germinal-center associated nuclear protein; germinal-centre associated nuclear protein; MCM3 acetylating protein; MCM3 acetyltransferase; MCM3 import protein; MCM3 minichromosome maintenance deficient 3 associated protein; MCM3AP
Gene Aliases: GANP; KIAA0572; MAP80; MCM3AP; SAC3
UniProt ID: (Human) O60318
Entrez Gene ID: (Human) 8888
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support