Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQRAGGGSAP GGNGGGGGGG PGTAFSIDSL IGPPPPRSGH LLYTGYPMFM PYRPLVLPQA LAPAPLPAGL PPLAPLASFA GRLTNTFCAG LGQAVPSMVA LTTALPSFAE PPDAFYGPQE LAAAAAAAAA TAARNNPEPG GRRPEGGLEA DELLPAREKV AEPPPPPPPH FSETFPSLPA EGKVYSSDEE KLEASAGDPA GSEQEEEGSG GDSEDDGFLD SSAGGPGALL GPKPKLKGSL GTGAEEGAPV TAGVTAPGGK SRRRRTAFTS EQLLELEKEF HCKKYLSLTE RSQIAHALKL SEVQVKIWFQ NRRAKWKRIK AGNVSSRSGE PVRNPKIVVP IPVHVNRFAV RSQHQQMEQG ARP
GBX1 gene ontology annotations related to this gene include adult walking behavior; neuron fate commitment; proprioception; sensory neuron axon guidance; spinal cord motor neuron differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gastrulation and brain-specific homeobox protein 1; Homeobox protein GBX-1
Gene Aliases: GBX1; Huh-17
UniProt ID: (Human) Q14549
Entrez Gene ID: (Human) 2636
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support