Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AVRGQGKDESK VEDDPKGKEE SFSLESDVDY SSDDNLTGQA AHKEEDPGHA LEET
Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.
Altered expression of GBX2, part of the homeobox-containing human family of DNA-binding transcription factors, is associated with therapy failure and death in patients with multiple types of cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gastrulation and brain-specific homeobox protein 2; Homeobox protein GBX-2
Gene Aliases: GBX2
UniProt ID: (Human) P52951
Entrez Gene ID: (Human) 2637
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support