Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive test controls include: THP-1, Mouse liver, Mouse brain, Mouse spleen, Rat lung, Rat liver. The target is usually found in the following locations: Cytoplasm, Nucleus.
Immunogen sequence: GERPRSEEDN ELNLPNLAAA YSSILSSLGE NPQRQGLLKT PWRAASAMQF FTKGYQETIS DVLNDAIFDE DHDEMVIVKD IDMFSMCEHH LVPFVGKVHI GYLPNKQVLG LSKLARIVEI YSRRLQVQER LTKQIAVAIT EALRPAGVGV VVEATHMCMV MRGVQKMNSK TVTSTMLGVF REDPKTREEF LTLIRS
This gene encodes a member of the GTP cyclohydrolase family. The encoded protein is the first and rate-limiting enzyme in tetrahydrobiopterin biosynthesis, catalyzing the conversion of GTP into 7,8-dihydroneopterin triphosphate. BH4 is an essential cofactor required by aromatic amino acid hydroxylases as well as nitric oxide synthases. Mutations in this gene are associated with malignant hyperphenylalaninemia and dopa-responsive dystonia. Several alternatively spliced transcript variants encoding different isoforms have been described; however, not all variants give rise to a functional enzyme.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dystonia 14; GTP cyclohydrolase 1; GTP cyclohydrolase I; GTP-CH-I; guanosine 5'-triphosphate cyclohydrolase I
Gene Aliases: DYT14; DYT5; DYT5a; GCH; GCH1; GTP-CH; GTP-CH-1; GTPCH; GTPCH1; HPABH4B
UniProt ID: (Human) P30793, (Mouse) Q05915, (Rat) P22288
Entrez Gene ID: (Human) 2643, (Mouse) 14528, (Rat) 29244
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support