Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MATPNNLTP TNCSWWPISA LESDAAKPAE APDAPEAASP AHWPRESLVL YHWTQSFSSQ KVRLVIAEKG LVCEERDVSL PQSEHKEPWF MRLNLGEEVP VIIHRDNIIS DYDQIIDYVE RTFTGEHVVA LMPEVGSLQH ARVLQYRELL DALPMDAYTH GCILHPELTT DSMIPKYATA EIRRHLANAT TDLMKLDHEE EPQLSEPYLS KQKKLMAKIL EHDDVSYLKK ILGELAMVLD QIEAELEKRK LENEGQKCEL WLCGCAFTLA DVLLGATLHR LKFLGLSKKY WEDGSRPNLQ SFFERVQRRF AFRKVLGDIH TTLLSAVIPN AFRLVKRKPP SFFGASFLMG SLGGMGYFAY WYLKKKYI (1-367 aa encoded by BC009014)
The ganglioside GD3 synthase causes cell differentiation with neurite sprouting when transfected into the mouse neuroblastoma cell line Neuro2a. After differentiation, the expression of several genes is upregulated, including one that encodes a protein termed ganglioside-induced differentiation-associated protein 1 (Gdap1). A similar gene was found in humans, and mutations in the human gene are associated with Charcot-Marie-Tooth type 4A disease. The protein encoded by this gene is similar in sequence to the human GDAP1 protein. Several transcript variants encoding different isoforms, as well as a noncoding transcript variant, have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ganglioside induced differentiation associated protein 1-like 1; Ganglioside-induced differentiation-associated protein 1-like 1; GDAP1-L1
Gene Aliases: dJ881L22.1; dJ995J12.1.1; Gdap1l; GDAP1L1
UniProt ID: (Human) Q96MZ0
Entrez Gene ID: (Human) 78997, (Mouse) 228858
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support