Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: CYRNVLQLA KEQSMSSVGF CVINSAKRGY PLEDATHIAL RTVRRFLEIH GETIEKVVFA VSDLEEGTYQ KLLPLYFPRS LKEENRSLPY LPADIGNAEG EPVVPERQIR ISEKPGAPED NQEEEDEGLG VDLSFIGSHA FARMEGDIDK QRKLILQGQL SEAALQKQHQ RNYNRWLCQA RSEDLSDIAS LKALYQTGVD NCGRTVMVVV GRNIPVTLID MDKALLYFIH VMDHIAVKEY VLVYFHTLTS EYNHLDSDFL KKLYDVVDVK YKRNLKAVYF VHPTFRSKVS TWFFTTFSVS GLKDKIHHVD SLHQLFSAIS PEQIDFPPFV LEYDARVRST RSSPSPGMVY (148-496 aa encoded by BC013132)
GDAP2 is a protein coding gene and gene ontology (GO) annotation includes lysosomal membrane; response to retinoic acid.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dJ776P7.1; Ganglioside-induced differentiation-associated protein 2; Ganglioside-induced differentiation-associated-protein 2
Gene Aliases: C77050; D3Ertd801e; GDAP2; MACROD3
UniProt ID: (Human) Q9NXN4, (Mouse) Q9DBL2, (Rat) Q66H63
Entrez Gene ID: (Human) 54834, (Mouse) 14547, (Rat) 362004
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support