Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: RMHADMMED VEEVYAGDIC ALFGIDCASG DTFTDKANSG LSMESIHVPD PVISIAMKPS NKNDLEKFSK GIGRFTREDP TFKVYFDTEN KETVISGMGE LHLEIYAQRL EREYGCPCIT GKPKVAFRET ITAPVPFDFT HKKQSGGAGQ YGKVIGVLEP LDPEDYTKLE FSDETFGSNI PKQFVPAVEK GFLDACEKGP LSGHKLSGLR FVLQDGAHHM VDSNEISFIR AGEGALKQAL ANATLCILEP IMAVEVVAPN EFQGQVIAGI NRRHGVITGQ DGVEDYFTLY ADVPLNDMFG YSTELRSCTE GKGEYTMEYS RYQPCLPSTQ EDVINKYLEA TGQLPVKKGK AKN (400-751 aa encoded by BC049210)
Eukaryotes contain two protein translational systems, one in the cytoplasm and one in the mitochondria. Mitochondrial translation is crucial for maintaining mitochondrial function and mutations in this system lead to a breakdown in the respiratory chain-oxidative phosphorylation system and to impaired maintenance of mitochondrial DNA. This gene encodes one of the mitochondrial translation elongation factors. Its role in the regulation of normal mitochondrial function and in different disease states attributed to mitochondrial dysfunction is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EF-Gmt; EF-Gmt {ECO:0000255|HAMAP-Rule:MF_03061}; Elongation factor G 1, mitochondrial; elongation factor G 1, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03061}; Elongation factor G, mitochondrial; elongation factor G, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03061}; Elongation factor G1; elongation factor G1 {ECO:0000255|HAMAP-Rule:MF_03061}; G elongation factor 1; G elongation factor, mitochondrial 1; G translation elongation factor, mitochondrial; GFM1; hEFG1; mEF-G 1; mEF-G 1 {ECO:0000255|HAMAP-Rule:MF_03061}; mitochondrial; mitochondrial elongation factor G; mitochondrial elongation factor G1
Gene Aliases: AW545374; COXPD1; D3Wsu133e; EF-G; EFG; EFG1; EFGM; EGF1; GFM; GFM1; hEFG1
UniProt ID: (Human) Q96RP9, (Mouse) Q8K0D5, (Rat) Q07803
Entrez Gene ID: (Human) 85476, (Mouse) 28030, (Rat) 114017
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support