Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
May be involved in spermatogenesis. Its mouse homolog is induced by dioxin and may play a role in spermatogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C3HC4-type zinc finger protein; DIF-3; dioxin inducible factor 3; Dioxin-inducible factor 3; Gametogenetin-binding protein 2; gametogenin binding protein 2; laryngeal carcinoma related gene 1; Laryngeal carcinoma-related protein 1; Protein ZNF403; zinc finger protein 403
Gene Aliases: AI451896; C75939; D330017P12Rik; DIF-3; DIF3; GGNBP2; LCRG1; LZK1; ZFP403; ZNF403
UniProt ID: (Human) Q9H3C7, (Rat) Q6GVH5, (Mouse) Q5SV77
Entrez Gene ID: (Human) 79893, (Rat) 360584, (Mouse) 217039
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support