Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGGFQRGKYG TMAEGRSEDN LSATPPALRI ILVGKTGCGK SATGNSILGQ PVFESKLRAQ SVTRTCQVKT GTWNGRKVLV VDTPSIFESQ ADTQELYKNI GDCYLLSAPG PHVLLLVIQL GRFTAQDTVA IRKVKEVFGT GAMRHVVILF THKEDLGGQA LDDYVANTDN CSLKDLVREC ERRYCAFNNW GSVEEQRQQQ AELLAVIERL GREREGSFHS NDLFLDAQLL QRTGAGACQE DYRQYQAKVE WQVEKHKQEL RENESNWAYK ALLRVKHLML LHYEIFVFLL LCSILFFIIF LFIFHYI
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTPase IMAP family member 5; IAN-5; immune associated nucleotide 4 like 1; Immune-associated nucleotide-binding protein 5; immunity associated protein 3; Immunity-associated nucleotide 4-like 1 protein; Immunity-associated nucleotide 5 protein; Immunity-associated protein 3; inhibitor of radiation- and OA-induced apoptosis, Irod/Ian5
Gene Aliases: GIMAP5; HIMAP3; IAN-5; IAN4; IAN4L1; IAN5; IMAP3; IROD
UniProt ID: (Human) Q96F15
Entrez Gene ID: (Human) 55340
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support