Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
GIPC1 is a scaffolding protein that regulates cell surface receptor expression and trafficking (Lee et al., 2008).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GAIP C-terminus-interacting protein; GIPC PDZ domain containing family, member 1; GLUT1 C-terminal binding protein; IGF-1 receptor interacting protein 1; PDZ domain-containing protein GIPC1; regulator of G-protein signalling 19 interacting protein 1; RGS-GAIP-interacting protein; RGS19-interacting protein 1; Synectin; Tax interaction protein 2; TIP-2
Gene Aliases: C19orf3; GIPC; GIPC1; GLUT1CBP; Hs.6454; IIP-1; NIP; RGS19IP1; SEMCAP; SYNECTIIN; SYNECTIN; TIP-2
UniProt ID: (Human) O14908
Entrez Gene ID: (Human) 10755
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support