Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASAVLSSVP TTASRFALLQ VDSGSGSDSE PGKGKGRNTG KSQTLGSKST TNEKKREKRR KKKEQQQSEA NELRNLAFKK IPQKSSHAVC NAQHDLPLSN PVQKDSREEN WQEWRQRDEQ LTSEMFEADL EKALLLSKLE YEEHKKEYED AENTSTQSKV MNKKDKRKNH QGKDRPLTVS LKDFHSEDHI SKKTEELSSS QTLSHDGGFF NRLEDDVHKI LIREKRREQL TEYNGTDNCT AHEHNQEVVL KDGRIERLKL ELERKDAEIQ KLKNVITQWE AKYKEVKARN AQLLKMLQEG EMKDKAEILL QVDESQSIKN ELTIQVTSLH AALEQERSKV KVLQAELAKY QGGRKGKRNS ESDQCR
The approximately 142 kDa Tankyrase protein is ubiquitously expressed in mammalian cells and is localized to human telomeres. It has homology to ankyrins and to the catalytic domain of poly (adenosine diphosphate-ribose) polymerase (PARP). Tankyrase binds to TRF1 (telomeric repeat binding factor-1), which negatively regulates the maintenance of telomere length.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cGMP-dependent protein kinase anchoring protein 42kDa; cGMP-dependent protein kinase-anchoring protein of 42 kDa; G kinase-anchoring protein 1; protein kinase anchoring protein GKAP42
Gene Aliases: FKSG21; GKAP1; GKAP42
UniProt ID: (Human) Q5VSY0
Entrez Gene ID: (Human) 80318
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support